missing translation for 'onlineSavingsMsg'
Learn More

Aurintricarboxylic acid

Product Code. 11478426
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478426 25 g 25g
11468426 5 g 5g
11488426 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478426 Supplier Thermo Scientific Alfa Aesar Supplier No. A15905.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4431-00-9 | C22H14O9 | 422.345 g/mol

Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Aurintricarboxylic acid is employed as a powerful inhibitor of cellular processes, including apoptosis, that are dependent on the formation of protein-nucleic acid complexes. It binds to a FGF and reduces its angiogenic activity. Aurintricarboxylic Acid has been shown to inhibit apoptotic cell death in various cell types induced by a variety of factors. It Inhibits protein-nucleic acid interactions. It stimulates the tyrosine phosphorylation of MAP kinases.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4431-00-9
Molecular Formula C22H14O9
Molecular Weight (g/mol) 422.345
MDL Number MFCD00011663
InChI Key GIXWDMTZECRIJT-UHFFFAOYSA-N
Synonym aurintricarboxylic acid, aluminon free acid, aluminon, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dienylidenemethylene di salicylic acid, benzoic acid, 5-3-carboxy-4-hydroxyphenyl 3-carboxy-4-oxo-2,5-cyclohexadien-1-ylidene methyl-2-hydroxy, 1,4-cyclohexadiene-1-carboxylic acid, 3-bis 3-carboxy-4-hydroxyphenyl methylene-6-oxo, 3,3'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 6-hydroxybenzoic acid, 5,5'-3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene methylene bis 2-hydroxybenzoic acid
PubChem CID 2259
ChEBI CHEBI:87397
IUPAC Name 5-[(3-carboxy-4-hydroxyphenyl)-(3-carboxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]-2-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1C(=C2C=CC(=O)C(=C2)C(=O)O)C3=CC(=C(C=C3)O)C(=O)O)C(=O)O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 2228904
Solubility Information Insoluble in water.
Formula Weight 422.35
Chemical Name or Material Aurintricarboxylic acid
compliance-icons
Product Identifier
  • Aurintricarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Aurintricarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.