Learn More
Hirschmann™ AR-Glass Graduated Pipet
Product Code. 11781516
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
25 mL
Graduations:
0.05 mL
0.1 mL
0.2 mL
Unit Size:
Pack of 12
Pack of 6
Product Code. Volume (Metric) Graduations unitSize
11781516 5 mL 0.05 mL Pack of 12
11716492 2 mL 0.1 mL Pack of 12
11726492 5 mL 0.1 mL Pack of 12
11736492 10 mL 0.1 mL Pack of 12
13127899 25 mL 0.1 mL Pack of 6
11746492 25 mL 0.2 mL Pack of 12
6 options
This item is not returnable. View return policy
Product Code. 11781516
Brand: Hirschmann™ 110015327
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Provides volumetric accuracy for your pipetting needs. Hirschmann™ Clear Glass Graduated Pipettes with Amber Stain Graduation with a cotton plug end as standard, short form for tissue culture.
  • Main point ring graduation
  • Cotton plug end
  • Class AS, Conformity certified, with dated batch
Specifications
Class A
Description Short form
Material Soda Lime Glass
Graduated Yes
Graduation Color Amber
Graduations 0.05 mL
Graduation Type Durable amber with silver diffusion color
Quantity 12 Pack
Tolerance 0.02 mL
Volume (Metric) 5 mL
Length (Metric) 360 ±5 mm
Type Graduated Pipet