Learn More
Antimony potassium tartrate hydrate, 98%
Product Code. 11444243
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
Product Code. Quantity unitSize
11444243 500 g 500g
11434243 100 g 100g
11454243 2500 g 2500g
3 options
This item is not returnable. View return policy
Product Code. 11444243
Brand: Thermo Scientific Alfa Aesar A10889.36
Request Bulk or Custom Format
This item is not returnable. View return policy
CAS: 331753-56-1 | C8H4K2O12Sb2 | 613.83 g/mol
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Antimony potassium tartrate hydrate is a compound used in the treatment of schistosomiasis. It is also used in the textile industry to aid in binding certain dyes to fabrics. It finds application in medicine as an expectorant and a nauseant.
Solubility
Soluble in water. Insoluble in ethanol.
Notes
Incompatible with mineral acids, strong bases, carbonates, lead, silver salts and strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 331753-56-1
Molecular Formula C8H4K2O12Sb2
Molecular Weight (g/mol) 613.83
MDL Number MFCD00148863
InChI Key GUJUCWZGYWASLH-UHFFFAOYNA-J
Synonym potassium antimony tartrate hydrate, potassium antimony iii tartrate hydrate, acmc-20ajv8, antimony 3+ potassium hydrate ditartrate
IUPAC Name dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
SMILES [K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
Specifications
Density 2.607
Assay Percent Range 98%
Quantity 500 g
UN Number UN1551
Solubility Information Soluble in water. Insoluble in ethanol.
Formula Weight 613.82 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Antimony potassium tartrate hydrate