missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD29 Hamster anti-Mouse, Unlabeled, Clone: HM Tests1-1, BD
Description
The HM β;1-1 monoclonal antibody specifically binds to the 130-kDa integrin β;1 chain (CD29). CD29 is expressed on the cell surface as a heterodimer with one of the distinct integrin-α; chains. With α;1 through α;6 (CD49a through CD49f), it forms the VLA-1 through VLA-6 complexes, respectively, and with α;v (CD51), it forms α;vβ;1 integrin. It also associates with the integrin α;7 α;8, and α;9 chains in non-lymphoid tissues. As a result, CD29 has a broad tissue distribution, including lymphocytes, endothelia, smooth muscle, epithelia, and oocytes. This hamster mAb to a mouse leukocyte antigen has been observed to crossreact with similar populations of rat leukocytes. Source of the immunogen was purified mouse VLA-4 (α;4β;1, CD49d/CD29).
Specifications
Specifications
| Antigen | CD29 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HM Tests1-1 |
| Concentration | 15.625μg/mL |
| Conjugate | Unconjugated |
| Description | Itgb1; Integrin beta-1; Fnrb; gpIIa; VLA-4 subunit beta |
| Formulation | Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. |
| Host Species | Hamster |
| Immunogen | Purified mouse VLA-4 |
| Show More |
For Research Use Only.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?