Learn More
Emma Safety Footwear Andes XD Safety Shoes
Product Code. 15918341
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
Product Code. Size unitSize
15918341 49 1 pair
15998321 37 1 pair
15908331 38 1 pair
15918331 39 1 pair
15928331 40 1 pair
15938331 41 1 pair
15948331 42 1 pair
15958331 43 1 pair
15968331 44 1 pair
15978331 45 1 pair
15988331 46 1 pair
15998331 47 1 pair
15908341 48 1 pair
13 options
This item is not returnable. View return policy
Product Code. 15918341
Brand: Emma Safety Footwear MM30556849
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Features a steel toe cap and an integrated stainless steel plate. The double-layer, anti-slip PU sole provides extra protection from hazards on the ground.
  • S3-Safety shoe with sole plate for wet work
  • Oil Nubuck construction
  • GripForce™ Easy Twist Outsole
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design Andes XD
Material Oiled Nubuck
Size 49