Descripción
Ideal for fishing, chemical processing, agriculture, petroleum refining, plant maintenance, heavy equipment assembly and cleanup
- Made from unsupported natural latex
- Offer abrasion, puncture, and tear resistance
- With extra heavy-duty, cotton, flock lining
- Length: 13 in. (33 cm)
- Thickness: 29 mil
- Orange
Especificaciones
Especificaciones
| Grado de abrasión ANSI | ANSI 3 |
| Certificaciones/conformidad | EN 374 AKL, EN 388 X121 |
| Certificaciones/conformidad (Europa) | CE 0493, EN 420:2003 + A1:2009; Category III |
| Chlorinated | Yes |
| Aplicaciones | Industrial |
| Color | Orange |
| Latex Free | No |
| Material | Latex |
| Palm Thickness (English) | 29.5 mil |
| Palm Thickness (Metric) | 0.75 mm |
| Mostrar más |