Learn More
Alendronate sodium trihydrate, 97%, Thermo Scientific™
51.60€ - 175.65€
Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15493999
SDS Ver documentos
Thermo Scientific Alfa Aesar
J61397.03
51.60€
1 g
Ver productos alternativos
Ahorre hasta 
»
15404009
SDS Ver documentos
Thermo Scientific Alfa Aesar
J61397.06
5g
ONLINE EXCLUSIVE PRICE
195.00€
175.65€ / 5 g
Ahorro 19.35€
9% Descuento
Ver productos alternativos
Ahorre hasta 
»
Descripción
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Alendronate sodium trihydrate is used as a bone resorption inhibitor, farnesyl diphosphate synthase inhibitor and CD45 protein tyrosine phosphatase inhibitor. A robust inhibitor of bone resorption that induces apoptosis. Alendronate inhibits the migration and invasion of PC-3 cells (sc-2220) by interfering with the mevalonate pathway. Additionally, Alendronate, Sodium Salt is an inhibitor of CD45RC and MMP.
Solubility
Soluble in water at 10mg/ml
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C. Keep away from strong oxidizing agents.