missing translation for 'onlineSavingsMsg'
Learn More

AEBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16808173 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16808173 150 μL 150µL
16818173 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16808173 Supplier Proteintech Supplier No. 112322AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AEBP2 is a DNA-binding transcriptional repressor. It may interact with and stimulate the activity of the PRC2 complex, which methylates ′Lys-9′ and ′Lys-27′ residues of histone H3.

Specifications

Antigen AEBP2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.26 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene AEBP2
Gene Accession No. Q6ZN18, Q9Z248
Gene Alias AE binding protein 2, AEBP2, Zinc finger protein AEBP2
Gene Symbols Aebp2
Host Species Rabbit
Immunogen AEBP2 Fusion Protein Ag1756
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11569, 121536, 297705
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.