Learn More
6-Bromo-7H-purine, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 767-69-1 |
| Molecular Formula | C5H3BrN4 |
| MDL Number | MFCD00022648 |
| Quantity | 1 g |
| SMILES | BrC1=NC=NC2=C1NC=N2 |
| Molecular Weight (g/mol) | 199.01 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Bromo-7H-purine |