Learn More
5-Oxotetrahydrofuran-2,3-dicarboxylic acid, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 4702-32-3 |
| Molecular Formula | C6H6O6 |
| MDL Number | MFCD00005397 |
| Quantity | 1 g |
| SMILES | O=C1OC(C(O)=O)C(C(O)=O)C1 |
| Molecular Weight (g/mol) | 174.11 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Oxotetrahydrofuran-2,3-dicarboxylic acid |