Learn More
5-Hydroxy-1-indanone, 97%, BLD Pharm™
Product Code. 30963972
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30963972 10 g 10g
30961231 25 g 25g
30960399 500 g 500g
30960086 5 g 5g
30954926 1 g 1g
30959888 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30963972 Supplier BLD Pharm Supplier No. BD1164410g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3470-49-3
Molecular Formula C9H8O2
MDL Number MFCD00857527
Quantity 10 g
SMILES C1=C(O)C=CC2=C1CCC2=O
Molecular Weight (g/mol) 148.16
Percent Purity 97%
Chemical Name or Material 5-Hydroxy-1-indanone