Learn More
5-Chloro-N-ethyl-4-hydroxy-1-methyl-2-oxo-N-phenyl-1,2-dihydroquinoline-3-carboxamide, 98%, BLD Pharm™
Product Code. 30946791
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
5 g
Unit Size:
100mg
1g
250mg
50mg
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30946791 50 mg 50mg
30947268 1 g 1g
30932355 5 g 5g
30937059 100 mg 100mg
30941994 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30946791 Supplier BLD Pharm Supplier No. BD12184350mg
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 248281-84-7
Molecular Formula C19H17ClN2O3
MDL Number MFCD08689859
Quantity 50 mg
SMILES C1=CC=C(Cl)C2=C1N(C(=C(C2=O)C(=O)N(CC)C3=CC=CC=C3)O)C
Molecular Weight (g/mol) 356.80
Percent Purity 98%
Chemical Name or Material 5-Chloro-N-ethyl-4-hydroxy-1-methyl-2-oxo-N-phenyl-1,2-dihydroquinoline-3-carboxamide