Learn More
5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulfamoyl)phenethyl)-2-methoxybenzamide, 98%, BLD Pharm™
Product Code. 31032707
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Unit Size:
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31032707 100 g -
31019860 25 g -
31018379 1 g -
31024469 500 g 500g
31027440 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31032707 Supplier BLD Pharm Supplier No. BD103656100g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 10238-21-8
Molecular Formula C23H28ClN3O5S
MDL Number MFCD00056625
Quantity 100 g
SMILES O=C(NCCC1=CC=C(S(=O)(NC(NC2CCCCC2)=O)=O)C=C1)C3=CC(Cl)=CC=C3OC
Molecular Weight (g/mol) 494.00
Percent Purity 98%
Chemical Name or Material 5-Chloro-N-(4-(N-(cyclohexylcarbamoyl)sulfamoyl)phenethyl)-2-methoxybenzamide