missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-hydrazino-3-iodopyridine, 95%, Thermo Scientific™

Product Code. 15423617
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
15423617 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 15423617

Brand: Thermo Scientific Alfa Aesar H27117.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 942206-12-4
Molecular Formula C5H5ClIN3
Molecular Weight (g/mol) 269.47
MDL Number MFCD09037446
InChI Key SMVJKHCZAOIBLS-UHFFFAOYSA-N
Synonym 5-chloro-3-iodo-2-pyridyl hydrazine, 5-chloro-2-hydrazinyl-3-iodopyridine, 5-chloro-2-hydrazino-3-iodopyridine
PubChem CID 42553001
IUPAC Name (5-chloro-3-iodopyridin-2-yl)hydrazine
SMILES C1=C(C=NC(=C1I)NN)Cl

Specifications

Melting Point 136°C to 138°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 269.47
Percent Purity 95%
Chemical Name or Material 5-Chloro-2-hydrazino-3-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.