Learn More
5-Chloro-2-fluorobenzaldehyde, 98%, BLD Pharm™
Product Code. 31340860
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
10g
25g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31340860 100 g -
31339615 500 g -
31339976 25 g 25g
31337947 1 g -
31338712 5 g 5g
31338189 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31340860 Supplier BLD Pharm Supplier No. BD10275100g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 96515-79-6
Molecular Formula C7H4ClFO
MDL Number MFCD03094518
Quantity 100 g
SMILES C1=C(C(=CC=C1Cl)F)C=O
Molecular Weight (g/mol) 158.56
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-fluorobenzaldehyde