Learn More
5-Bromopentyl acetate, 95%, BLD Pharm™
Product Code. 30774007
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30774007 500 g 500g
30771596 5 g 5g
30772398 100 g 100g
30773599 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30774007 Supplier BLD Pharm Supplier No. BD103104500g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 15848-22-3
Molecular Formula C7H13BrO2
MDL Number MFCD00039204
Quantity 500 g
SMILES CC(OCCCCCBr)=O
Molecular Weight (g/mol) 209.08
Percent Purity 95%
Chemical Name or Material 5-Bromopentyl acetate