Learn More
5,9-Diaminobenzo[a]phenoxazin-7-ium acetate, 65%, BLD Pharm™
Specifications
Specifications
| CAS | 10510-54-0 |
| Molecular Formula | C18H15N3O3 |
| MDL Number | MFCD00013151 |
| Quantity | 10 g |
| SMILES | NC1=CC2=[O+]C3=C(C4=CC=CC=C4C(N)=C3)N=C2C=C1.CC([O-])=O |
| Molecular Weight (g/mol) | 321.33 |
| Percent Purity | 65% |
| Chemical Name or Material | 5,9-Diaminobenzo[a]phenoxazin-7-ium acetate |