Learn More
5,5'-Dibromo-2,2'-bithiophene, 98%, BLD Pharm™
Product Code. 31006647
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31006647 5 g 5g
31004572 25 g 25g
30996917 10 g 10g
31001108 500 g 500g
30991282 100 g 100g
31003984 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31006647 Supplier BLD Pharm Supplier No. BD108135g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 4805-22-5
Molecular Formula C8H4Br2S2
MDL Number MFCD00219110
Quantity 5 g
SMILES BrC1=CC=C(S1)C2=CC=C(S2)Br
Molecular Weight (g/mol) 324.06
Percent Purity 98%
Chemical Name or Material 5,5'-Dibromo-2,2'-bithiophene