missing translation for 'onlineSavingsMsg'
Learn More

4-(Pyrrolidin-1-ylmethyl)benzaldehyde, ≥97%, Thermo Scientific™

Product Code. 10074243
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10074243 1 g 1g
10527474 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10074243 Supplier Thermo Scientific Maybridge Supplier No. CC55604DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 650628-72-1
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD07782421
InChI Key MTBYCACQSYAYCC-UHFFFAOYSA-N
PubChem CID 23090828
IUPAC Name 4-(pyrrolidin-1-ylmethyl)benzaldehyde
SMILES C1CCN(C1)CC2=CC=C(C=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Synonym 4-pyrrolidin-1-ylmethyl benzaldehyde, 4-1-pyrrolidinylmethyl benzaldehyde, benzaldehyde, 4-1-pyrrolidinylmethyl, 4-pyrrolidin-1-yl methyl-benzaldehyde, 1-4-formylbenzyl pyrrolidine, 4-pyrrolizinomethyl benzaldehyde, 4-pyrrolidinylmethyl benzaldehyde, 4-pyrrolidin-1-ylmethyl-benzaldehyde, benzaldehyde,4-1-pyrrolidinylmethyl, 4-pyrrolidin-1-yl methyl benzaldehyde
Formula Weight 189.26
Percent Purity 97%
Chemical Name or Material 4-(Pyrrolidin-1-ylmethyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.