missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenoxymethyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10092014
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10092014

Brand: Thermo Scientific Maybridge CC63701DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 31719-76-3
Molecular Formula C14H11O3
Molecular Weight (g/mol) 227.24
MDL Number MFCD00623268
InChI Key GRBUVHSYBRTCIB-UHFFFAOYSA-M
Synonym 4-phenoxymethyl benzoic acid, 4-phenoxymethyl benzenecarboxylic acid, 4-phenoxymethyl-benzoic acid, phenoxymethylbenzenecarboxylicacid
PubChem CID 737267
IUPAC Name 4-(phenoxymethyl)benzoic acid
SMILES [O-]C(=O)C1=CC=C(COC2=CC=CC=C2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 228.25
Percent Purity 97%
Chemical Name or Material 4-(Phenoxymethyl)benzoic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.