Learn More
4-Nitronaphthalen-1-amine, 97%, BLD Pharm™
Product Code. 31450250
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31450250 500 g
31434918 5 g
31441610 10 g
31441736 25 g
31448790 1 g
31433507 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 31450250 Supplier BLD Pharm Supplier No. BD12726500g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 776-34-1
Molecular Formula C10H8N2O2
MDL Number MFCD00004026
Quantity 500 g
SMILES NC1=CC=C(C2=C1C=CC=C2)[N+]([O-])=O
Molecular Weight (g/mol) 188.18
Percent Purity 97%
Chemical Name or Material 4-Nitronaphthalen-1-amine