Learn More
(4-(Morpholine-4-carbonyl)phenyl)boronic acid, 98%, BLD Pharm™
Product Code. 30986135
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30986135 5 g -
30971859 1 g -
30980143 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30986135 Supplier BLD Pharm Supplier No. BD132175g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 389621-84-5
Molecular Formula C11H14BNO4
MDL Number MFCD03411952
Quantity 5 g
SMILES O=C(C1=CC=C(B(O)O)C=C1)N2CCOCC2
Molecular Weight (g/mol) 235.04
Percent Purity 98%
Chemical Name or Material (4-(Morpholine-4-carbonyl)phenyl)boronic acid