Learn More
4-(Imidazol-1-yl)phenol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 10041-02-8 |
| Molecular Formula | C9H8N2O |
| MDL Number | MFCD00005281 |
| Quantity | 5 g |
| SMILES | OC1=CC=C(N2C=CN=C2)C=C1 |
| Molecular Weight (g/mol) | 160.17 |
| Percent Purity | 95% |
| Chemical Name or Material | 4-(Imidazol-1-yl)phenol |