Learn More
4-Chloromandelic acid, 98%, BLD Pharm™
Product Code. 31001694
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31001694 100 g 100g
30995914 1 kg -
30992800 5 g 5g
30993847 25 g 25g
30995689 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31001694 Supplier BLD Pharm Supplier No. BD11034100g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 492-86-4
Molecular Formula C8H7ClO3
MDL Number MFCD00042724
Quantity 100 g
SMILES OC(C(O)=O)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 186.59
Percent Purity 98%
Chemical Name or Material 4-Chloromandelic acid