Learn More
4-(4-Methoxyphenyl)-4-oxobutanoic acid, 98%, BLD Pharm™
Product Code. 30964987
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
1g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30964987 5 g 5g
30958986 10 g 10g
30959636 25 g 25g
30962199 1 g 1g
30957029 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30964987 Supplier BLD Pharm Supplier No. BD131435g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 3153-44-4
Molecular Formula C11H12O4
MDL Number MFCD00002795
Quantity 5 g
SMILES O=C(O)CCC(C1=CC=C(OC)C=C1)=O
Molecular Weight (g/mol) 208.21
Percent Purity 98%
Chemical Name or Material 4-(4-Methoxyphenyl)-4-oxobutanoic acid