missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dimethyl-2,2'-bipyridine, 98%

Product Code. 11471447
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. Quantity unitSize
11471447 5 g 5g
11461447 1 g 1g
11481447 25 g 25g
3 options
This item is not returnable. View return policy

Product Code. 11471447

Brand: Thermo Scientific Alfa Aesar B24936.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1134-35-6 | C12H12N2 | 184.242 g/mol

4,4'-Dimethyl-2,2'-bipyridine is used as a chemical Intermediate. It can be used for the determination of ferrous and cyanide compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dimethyl-2,2′-bipyridine is used as a chemical Intermediate. It can be used for the determination of ferrous and cyanide compounds.

Solubility
Soluble in ethanol, acetic acid, benzene, toluene. Highly soluble in water at pH < 2.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1134-35-6
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00006441
InChI Key NBPGPQJFYXNFKN-UHFFFAOYSA-N
Synonym 4,4'-dimethyl-2,2'-bipyridine, 4,4'-dimethyl-2,2'-bipyridyl, 4,4'-dimethyl-2,2'-dipyridyl, 2,2'-bi-4-picoline, 2,2'-bipyridine, 4,4'-dimethyl, 2,2'-bi gamma-picoline, 2,2'-di-4-picolyl, 4-methyl-2-4-methylpyridin-2-yl pyridine, 2,2'-bipyridyl, 4,4'-dimethyl, 4,4-dimethyl-2,2-bipyridyl
PubChem CID 14338
IUPAC Name 4-methyl-2-(4-methylpyridin-2-yl)pyridine
SMILES CC1=CC(=NC=C1)C2=NC=CC(=C2)C

Specifications

Melting Point 171°C to 175°C
Quantity 5 g
Beilstein 128995
Solubility Information Soluble in ethanol,acetic acid,benzene,toluene. Highly soluble in water at pH < 2.
Formula Weight 184.24
Percent Purity 98%
Chemical Name or Material 4,4'-Dimethyl-2,2'-bipyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.