Learn More
4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 171364-83-3 |
| Molecular Formula | C12H16BNO4 |
| MDL Number | MFCD02179437 |
| Quantity | 25 g |
| SMILES | O=[N+](C1=CC=C(B(OC2(C)C)OC2(C)C)C=C1)[O-] |
| Molecular Weight (g/mol) | 249.07 |
| Percent Purity | 98% |
| Chemical Name or Material | 4,4,5,5-Tetramethyl-2-(4-nitrophenyl)-1,3,2-dioxaborolane |