missing translation for 'onlineSavingsMsg'
Learn More

4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine, Thermo Scientific Chemicals

Product Code. 11474790
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11474790 250 mg 250mg
11484790 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 11474790

Brand: Thermo Scientific Alfa Aesar H50804.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-38-8
Molecular Formula C14H16ClN3
Molecular Weight (g/mol) 261.75
MDL Number MFCD08060997
InChI Key QPXZSTSEYCYKLI-UHFFFAOYSA-N
Synonym 4-4-4-chlorophenyl-1h-pyrazol-1-yl piperidine, 4-4-4-chlorophenyl pyrazol-1-yl piperidine, acmc-20amql, 4-4-chlorophenyl-1-4-piperidyl pyrazole, 4-4-4-chlorophenyl-1h-pyrazol-1-ylpiperidine, 4-4-4-chloro-phenyl-pyrazol-1-yl-piperidine
PubChem CID 24212026
IUPAC Name 4-[4-(4-chlorophenyl)pyrazol-1-yl]piperidine
SMILES ClC1=CC=C(C=C1)C1=CN(N=C1)C1CCNCC1

Specifications

Quantity 250 mg
Formula Weight 261.76
Chemical Name or Material 4-[4-(4-Chlorophenyl)-1H-pyrazol-1-yl]piperidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.