missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Product Code. 15428515
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428515 250 mg 250mg
15406347 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428515 Supplier Thermo Scientific Alfa Aesar Supplier No. H33520.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
A series of novel five-membered urea derivatives as potent NK1 receptor antagonists is described. The effects of substitution of a 4-fluoro group at the phenyl ring and the introduction of an α-methyl group at the benzylic position to improve potency was found. 3,5-Bis(trifluoromethyl)benzyl bromide has been used as derivatization reagent in detection of uracil in DNA by GC and negative chemical ionization mass spectrometry.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116070-36-1
Molecular Formula C9H8BrF3
Molecular Weight (g/mol) 253.06
MDL Number MFCD01631610
InChI Key GZJBZDJGNUXMNN-UHFFFAOYSA-N
Synonym 1-bromomethyl-3-methyl-5-trifluoromethyl benzene, 3-methyl-5-trifluoromethyl benzyl bromide, 3-trifluoromethvl-5-methvlbenzvl bromide, 3-trifluoromethyl-5-methylbenzyl bromide, benzene, 1-bromomethyl-3-methyl-5-trifluoromethyl
PubChem CID 2775623
IUPAC Name 1-(bromomethyl)-3-methyl-5-(trifluoromethyl)benzene
SMILES CC1=CC(CBr)=CC(=C1)C(F)(F)F

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 253.06
Percent Purity 97%
Chemical Name or Material 3-Methyl-5-(trifluoromethyl)benzyl bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.