Learn More
3-Isopropyl 5-methyl 4-(benzo[c][1,2,5]oxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 98%, BLD Pharm™
Product Code. 31448367
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31448367 5 g
31440015 250 mg
31440379 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31448367 Supplier BLD Pharm Supplier No. BD1354675g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 75695-93-1
Molecular Formula C19H21N3O5
MDL Number MFCD00153820
Quantity 5 g
SMILES O=C(C1=C(C)NC(C)=C(C(OC(C)C)=O)C1C2=CC=CC3=NON=C32)OC
Molecular Weight (g/mol) 371.39
Percent Purity 98%
Chemical Name or Material 3-Isopropyl 5-methyl 4-(benzo[c][1,2,5]oxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate