missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylphenylacetic acid, 97%, Thermo Scientific™

Product Code. 11332629
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
11332629 1 g 1g
15423527 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 11332629

Brand: Thermo Scientific Alfa Aesar H26690.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261951-74-0
Molecular Formula C9H9FO2
Molecular Weight (g/mol) 168.17
MDL Number MFCD01631536
InChI Key YECYDEGFKJWADM-UHFFFAOYSA-N
Synonym 3-fluoro-4-methylphenylacetic acid, 2-3-fluoro-4-methylphenyl acetic acid, 3-fluoro-4-methylphenyl acetic acid, benzeneacetic acid, 3-fluoro-4-methyl, 3-fluoro-4-methylphenylaceticacid, acmc-20aica, pubchem5864, 3-fluoro-4-methyl-phenyl-acetic acid
PubChem CID 2774618
IUPAC Name 2-(3-fluoro-4-methylphenyl)acetic acid
SMILES CC1=C(F)C=C(CC(O)=O)C=C1

Specifications

Melting Point 104°C to 107°C
Quantity 1 g
Formula Weight 168.17
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylphenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.