missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3-Aminopiperidine dihydrochloride, 95%, Thermo Scientific™

Product Code. 15423818
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423818 250 mg 250mg
15413818 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423818 Supplier Thermo Scientific Alfa Aesar Supplier No. H57306.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as a reactant for N-arylation of heterocyclic diamines and for synthesis of substituted quinolones with reduced phototoxic risk.

Solubility
Partly miscible in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138060-07-8
Molecular Formula C5H14Cl2N2
Molecular Weight (g/mol) 173.081
MDL Number MFCD00012773
InChI Key GGPNYXIOFZLNKW-UHFFFAOYSA-N
Synonym 3-aminopiperidine dihydrochloride, piperidin-3-amine dihydrochloride, 3-aminopiperidine 2hcl, 3-aminopiperidine hcl, 3-piperidinamine dihydrochloride, 3-piperidinamine, dihydrochloride, pubchem6755, acmc-209tvl, ksc839o8j, 3-amino-piperidine di-hydrochloride
PubChem CID 16211870
IUPAC Name piperidin-3-amine;dihydrochloride
SMILES C1CC(CNC1)N.Cl.Cl

Specifications

Melting Point 230°C
Quantity 250 mg
Sensitivity Hygroscopic
Solubility Information Partly miscible in water.
Formula Weight 173.09
Percent Purity 95%
Chemical Name or Material (+/-)-3-Aminopiperidine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.