Learn More
3-Amino-7-(methylamino)phenothiazin-5-ium chloride, 40%, BLD Pharm™
Product Code. 30999687
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30999687 5 g 5g
30992575 1 g 1g
30995624 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30999687 Supplier BLD Pharm Supplier No. BD1149985g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 531-57-7
Molecular Formula C13H12ClN3S
MDL Number MFCD00067677
Quantity 5 g
SMILES [Cl-].CNC1=CC2=C(C=C1)N=C1C=CC(N)=CC1=[S+]2
Molecular Weight (g/mol) 277.77
Percent Purity 40%
Chemical Name or Material 3-Amino-7-(methylamino)phenothiazin-5-ium chloride