Learn More
3-Amino-7-(diethylamino)-5-phenylphenazin-5-ium chloride, 98%, BLD Pharm™
Product Code. 31004198
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31004198 5 g 5g
30998796 25 g 25g
30999725 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31004198 Supplier BLD Pharm Supplier No. BD1212365g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 4569-86-2
Molecular Formula C22H23ClN4
MDL Number MFCD00013433
Quantity 5 g
SMILES CCN(C1=CC2=[N+](C3=CC=CC=C3)C4=C(C=CC(N)=C4)N=C2C=C1)CC.[Cl-]
Molecular Weight (g/mol) 378.90
Percent Purity 98%
Chemical Name or Material 3-Amino-7-(diethylamino)-5-phenylphenazin-5-ium chloride