Learn More
3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide, 95%, BLD Pharm™
Product Code. 30948981
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
25mg
50mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30948981 50 mg 50mg
30942615 1 g 1g
30938979 100 mg 100mg
30946559 25 mg 25mg
30939810 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30948981 Supplier BLD Pharm Supplier No. BD14923450mg
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 25535-16-4
Molecular Formula C27H34I2N4
MDL Number MFCD00011921
Quantity 50 mg
SMILES C[N+](CCC[N+]1=C(C2=CC=CC=C2)C3=CC(N)=CC=C3C4=C1C=C(N)C=C4)(CC)CC.[I-].[I-]
Molecular Weight (g/mol) 668.39
Percent Purity 95%
Chemical Name or Material 3,8-Diamino-5-(3-(diethyl(methyl)ammonio)propyl)-6-phenylphenanthridin-5-ium iodide