Learn More
3,3-Pentamethylene glutarimide, 97%, BLD Pharm™
Product Code. 31029460
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
500 g
Unit Size:
500g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31029460 100 g -
31022141 500 g 500g
31022669 25 g -
31016088 5 g -
31018625 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 31029460 Supplier BLD Pharm Supplier No. BD12296100g
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 1130-32-1
Molecular Formula C10H15NO2
MDL Number MFCD00023872
Quantity 100 g
SMILES O=C1CC2(CC(=O)N1)CCCCC2
Molecular Weight (g/mol) 181.23
Percent Purity 97%
Chemical Name or Material 3,3-Pentamethylene glutarimide