Learn More
3-(2-(Benzyl(methyl)amino)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 54527-84-3 |
| Molecular Formula | C26H30ClN3O6 |
| MDL Number | MFCD00057327 |
| Quantity | 5 g |
| SMILES | O=C(C1=C(C)NC(C)=C(C(OCCN(C)CC2=CC=CC=C2)=O)C1C3=CC=CC([N+]([O-])=O)=C3)OC.[H]Cl |
| Molecular Weight (g/mol) | 515.99 |
| Percent Purity | 99% |
| Chemical Name or Material | 3-(2-(Benzyl(methyl)amino)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate hydrochloride |