missing translation for 'onlineSavingsMsg'
Learn More

2-piperidinoaniline, 97%, Thermo Scientific™

Product Code. 10193953
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10193953 10 g 10g
10143064 1 g 1g
10693985 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10193953 Supplier Thermo Scientific Maybridge Supplier No. BTB02217EA
 Restricted Item
Fecha estimada de envoi: 15-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 39643-31-7
Molecular Formula C11H16N2
Molecular Weight (g/mol) 176.26
MDL Number MFCD00047467
InChI Key OYECAJPUPWFCSL-UHFFFAOYSA-N
Synonym 2-piperidinoaniline, 2-piperidin-1-yl aniline, 2-piperidin-1-yl-phenylamine, benzenamine, 2-1-piperidinyl, n-2-aminophenyl piperidine, 2-1-piperidinyl aniline, 2-1-piperidino aniline, 1-2-aminophenyl piperidine, 2-piperidylphenylamine, 2-piperdinoaniline
PubChem CID 458795
IUPAC Name 2-piperidin-1-ylaniline
SMILES NC1=CC=CC=C1N1CCCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 176.26
Percent Purity 97%
Chemical Name or Material 2-Piperidinoaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.