missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentenoic acid, 99%

Product Code. 11477917
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11477917 50 g 50g
11467917 10 g 10g
11487917 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477917 Supplier Thermo Scientific Alfa Aesar Supplier No. A16587.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3142-72-1 | C6H10O2 | 114.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3142-72-1
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00002655
InChI Key JJYWRQLLQAKNAD-SNAWJCMRSA-N
Synonym 2-methyl-2-pentenoic acid, trans-2-methyl-2-pentenoic acid, e-2-methylpent-2-enoic acid, e-2-methyl-2-pentenoic acid, 2-pentenoic acid, 2-methyl, 2-pentenoic acid, 2-methyl-, e, e-2-methylpent-2-en-1-oic acid, s-2-methylpentenoic acid, 2e-2-methylpent-2-enoic acid, 2-methylpent-2-en-1-oic acid
PubChem CID 5365909
IUPAC Name (E)-2-methylpent-2-enoic acid
SMILES CCC=C(C)C(=O)O

Specifications

Density 0.978
Boiling Point 120°C (20 mmHg)
Flash Point 107°C (225°F)
Odor Characteristic
Quantity 50 g
UN Number UN3265
Formula Weight 114.15
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-pentenoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.