Learn More
2-Methoxy-4-nitrobenzoic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2597-56-0 |
| Molecular Formula | C8H7NO5 |
| MDL Number | MFCD00216573 |
| Quantity | 25 g |
| SMILES | COC1=C(C=CC(=C1)[N+]([O-])=O)C(O)=O |
| Molecular Weight (g/mol) | 197.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Methoxy-4-nitrobenzoic acid |