Learn More
2-Mercapto-6-phenylpyrimidin-4-ol, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 36822-11-4 |
| Molecular Formula | C10H8N2OS |
| MDL Number | MFCD00059166 |
| Quantity | 10 g |
| SMILES | OC1=NC(S)=NC(C2=CC=CC=C2)=C1 |
| Molecular Weight (g/mol) | 204.25 |
| Percent Purity | 95% |
| Chemical Name or Material | 2-Mercapto-6-phenylpyrimidin-4-ol |