Learn More
2'-Fluoro-4'-methylacetophenone, 97%, Thermo Scientific™
Product Code. 11312639
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11312639 1 g 1g
11322639 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312639 Supplier Thermo Scientific Alfa Aesar Supplier No. H26695.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
2′-Fluoro-4′-methylacetophenone is used as an intermediate in pharmaceuticals.
Notes
Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29427-48-3
Molecular Formula C9H9FO
Molecular Weight (g/mol) 152.17
MDL Number MFCD03094331
InChI Key IBABHBXTLZNGAV-UHFFFAOYSA-N
Synonym 1-2-fluoro-4-methylphenyl ethanone, 2'-fluoro-4'-methylacetophenone, 1-2-fluoro-4-methylphenyl ethan-1-one, ethanone, 1-2-fluoro-4-methylphenyl, acmc-1cjnp, 4-acetyl-3-fluorotoluene, 2-fluoro-4-methylacetophenone, 1-acetyl-2-fluoro-4-methylbenzene, ethanone,1-2-fluoro-4-methylphenyl
PubChem CID 2778484
IUPAC Name 1-(2-fluoro-4-methylphenyl)ethanone
SMILES CC(=O)C1=C(F)C=C(C)C=C1
Specifications
Quantity 1 g
Formula Weight 152.17
Percent Purity 97%
Chemical Name or Material 2'-Fluoro-4'-methylacetophenone