Learn More
2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid hydrochloride, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 13558-31-1 |
| Molecular Formula | C20H15ClN2O3 |
| MDL Number | MFCD00042009 |
| Quantity | 5 g |
| SMILES | O=C(O)C1=CC=CC=C1C2=C3C=CC(C=C3OC4=C2C=CC(N)=C4)=N.[H]Cl |
| Molecular Weight (g/mol) | 366.80 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-(6-Amino-3-imino-3H-xanthen-9-yl)benzoic acid hydrochloride |