Learn More
2,5-Dioxopyrrolidin-1-yl 7-methoxy-2-oxo-2H-chromene-3-carboxylate, 98%, BLD Pharm™
Product Code. 30783123
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30783123 100 mg 100mg
30770647 250 mg 250mg
30782085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30783123 Supplier BLD Pharm Supplier No. BD145692100mg
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 150321-92-9
Molecular Formula C15H11NO7
MDL Number MFCD00467593
Quantity 100 mg
SMILES COC1=CC=C2C=C(C(=O)ON3C(=O)CCC3=O)C(=O)OC2=C1
Molecular Weight (g/mol) 317.25
Percent Purity 98%
Chemical Name or Material 2,5-Dioxopyrrolidin-1-yl 7-methoxy-2-oxo-2H-chromene-3-carboxylate