Learn More
((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hemisulfate, 99%, BLD Pharm™
Product Code. 30799525
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30799525 100 mg 100mg
30786609 250 mg 250mg
30784804 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30799525 Supplier BLD Pharm Supplier No. BD111900100mg
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 188062-50-2
Molecular Formula C28H38N12O6S
MDL Number MFCD04112763
Quantity 100 mg
SMILES NC1=NC(NC2CC2)=C3N=CN([C@H]4C=C[C@@H](CO)C4)C3=N1.O=S(O)(O)=O.NC5=NC(NC6CC6)=C7N=CN([C@H]8C=C[C@@H](CO)C8)C7=N5
Molecular Weight (g/mol) 670.74
Percent Purity 99%
Chemical Name or Material ((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol hemisulfate