Learn More
Melford 17-AAG
Product Code. 30017966
Change view
Click to view available options
Quantity:
1 mg
500 μg
Unit Size:
1mg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30017966 1 mg 1mg
30017965 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30017966 Supplier Melford Supplier No. A200450.001
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
17-AAG (17-Allylamino-17-demethoxygeldanamycin) is a synthetic derivative of geldanamycin. Like geldanamycin, 17-AAG is recognized for its role as an inhibitor of heat shock protein 90 (Hsp90), a molecular chaperone involved in protein folding and stabilization.
17-AAG (17-Allylamino-17-demethoxygeldanamycin) is a synthetic derivative of geldanamycin. Like geldanamycin, 17-AAG is recognized for its role as an inhibitor of heat shock protein 90 (Hsp90), a molecular chaperone involved in protein folding and stabilization.
It is often used as a tool to study Hsp90-dependent processes. By inhibiting Hsp90, 17-AAG impacts the stability and function of various client proteins, many of which are involved in critical cellular processes, including signal transduction and cell cycle regulation. Similar to geldanamycin, 17-AAG is particularly relevant in cancer research, as Hsp90 is often crucial for the stability of oncoproteins.
Researchers use 17-AAG to explore the molecular mechanisms of Hsp90 inhibition, with potential applications in understanding cancer biology and developing therapeutic strategies.
Specifications
CAS 75747-14-7
Molecular Formula C31H43N3O8
Quantity 1 mg