Learn More
R&D Systems™ 10058-F4
Product Code. 30001996
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001996 50 mg 50mg
30001987 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001996 Supplier R&D Systems™ Supplier No. 4406/50
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Tocris product, part of R&D Systems. Inhibits c-Myc-Max dimerization
10058-F4 is a cell permeable c-Myc-Max dimerization inhibitor. Inhibits proliferation, induces apoptosis and arrests cells in G0/G1 in rat1a-c-Myc cells. Also reduces tumor growth in vivo. In lymphoma cells, 10058-F4 decreases glucose uptake and expression of glycolysis-associated genes and reduces cell viability.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 403811-55-2
Molecular Formula C12H11NOS2
Specifications
Quantity 50 mg
Product Line Tocris
Formula Weight 249.35
Physical Form Solid
Color Yellow
Purity >99%
Molecular Weight (g/mol) 249.346
Chemical Name or Material 10058-F4