Learn More
1-Tosyl-1H-pyrrole, 98%, BLD Pharm™
Product Code. 30801879
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
1g
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30801879 100 g 100g
30787616 5 g 5g
30795877 25 g 25g
30789582 500 g 500g
30800813 1 g 1g
30794650 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 30801879 Supplier BLD Pharm Supplier No. BD13030100g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 17639-64-4
Molecular Formula C11H11NO2S
MDL Number MFCD00145014
Quantity 100 g
SMILES O=S(N1C=CC=C1)(C2=CC=C(C)C=C2)=O
Molecular Weight (g/mol) 221.28
Percent Purity 98%
Chemical Name or Material 1-Tosyl-1H-pyrrole