Learn More
1,6-Dibromonaphthalen-2-ol, 97%, BLD Pharm™
Product Code. 30783519
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
Unit Size:
100g
10g
25g
500g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30783519 10 g 10g
30781989 500 g 500g
30777518 5 g 5g
30768576 100 g 100g
30774766 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30783519 Supplier BLD Pharm Supplier No. BD1249510g
 Restricted Item
Fecha estimada de envoi: 25-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Specifications
CAS 16239-18-2
Molecular Formula C10H6Br2O
MDL Number MFCD00003870
Quantity 10 g
SMILES OC1=CC=C2C=C(Br)C=CC2=C1Br
Molecular Weight (g/mol) 301.96
Percent Purity 97%
Chemical Name or Material 1,6-Dibromonaphthalen-2-ol