Learn More
1,4-Dioxan-2-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 3041-16-5 |
| Molecular Formula | C4H6O3 |
| MDL Number | MFCD00215855 |
| Quantity | 5 g |
| SMILES | O=C1OCCOC1 |
| Molecular Weight (g/mol) | 102.09 |
| Percent Purity | 98% |
| Chemical Name or Material | 1,4-Dioxan-2-one |